Learn More
Biospec Products Mini-BeadBeater Zirconia-Silicate Beads
Product Code. 13468236 Shop All Cole-Parmer Products
Change view
Click to view available options
Quantity:
450 g (1 lb.)
Unit Size:
450g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13468236 450 g (1 lb.) 450g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13468236 Supplier Biospec Products Supplier No. 3627062
  • ONLINE EXCLUSIVE PRICE
    1880.00 NOK
    1164.02 NOK / 450g
    Save 715.98 NOK
    38% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Mini-BeadBeater Zirconia-Silicate Beads to suit your disruption application needs.
  • 1 year warranty.
Specifications
Type Beads
Material Zirconia-Silicate
Diameter (Metric) 0.1 to 0.15 mm
For Use With (Equipment) Mini-Bead Beater Glass Mill
Quantity 450 g (1 lb.)