Learn More
Oakton™ Glass Beads for Mills
Product Code. 13478236
Change view
Click to view available options
Diameter (Metric):
0.1 to 0.15 mm
0.5 mm
1 mm
Unit Size:
1kg
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
13478236 0.1 to 0.15 mm 1kg
15237649 1 mm Each
12614267 0.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13478236 Supplier Oakton™ Supplier No. 3627052
Estimated Shipment: 29-12-2026
Suitable glass beads for the mini-BeedBeater™ Mill. Cole-Parmer™ Glass Beads for Mills are used to crush and separate small samples from bone to microbial spores.
  • Available in three different sizes
  • Crush samples such as bone and microbial spores
  • Handles small samples efficiently—from 100μL to 1μL
  • Non-foaming, non-aerosol method preserves enzymes, nucleic acids, and organelles
Specifications
Type Beads
Material Glass
Diameter (Metric) 0.1 to 0.15 mm
For Use With (Equipment) Mini-Bead Beater Glass Mill
Quantity 1 lb.