missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIF Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00148AF700
This item is not returnable.
View return policy
Description
GIF Polyclonal specifically detects GIF in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GIF | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| gastric intrinsic factor (vitamin B synthesis), IFIntrinsic factor, IFMHgastric intrinsic factor, INF, TCN3 | |
| Produced in rabbits immunized with purified, recombinant GIF (P27352-1; Met1-Tyr417). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 2694 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction