Learn More
Gibco™ Fetal Bovine Serum, dialyzed , US Origin
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Specifications
Specifications
| Format | Bottle |
| Quantity | 100 mL |
| Endotoxin Concentration | ≤50 EU/mL (Specification is checked and recorded; levels routinely ≤ 10 EU/mL) |
| Heat Inactivated | No |