Learn More
Gibberellic Acid, Sigma-Aldrich™
Specifications
Specifications
| CAS | 77-06-5 |
| Melting Point | 233°C to 235°C |
| Density | 1.34 g/cm3 at 20°C |
| MDL Number | MFCD00079329 |
| Quantity | 1 g |
| Synonym | Gibberellic acid, Gibberellic acid |
| Molecular Weight (g/mol) | 346.37 |
| Grade | Research |