Learn More
Brady™ GHS Symbol - Oxidizing, Continuous Roll
Click to view available options
Adhesive Type:
Permanent Acrylic
Unit Size:
Pack of 250
Description
- High-quality labels that can withstand the most demanding environments
- Highly resistant to bad weather, abrasion and chemical products
Specifications
Specifications
| Alternate Part Number | PIC 1803-100*100-B7541-RLL-C |
| Finish | Gloss |
| Material | Polyester |
| Quantity | 250/Pk. |
| Adhesive Type | Permanent Acrylic |
| Height (Metric) | 100 mm |
| Thickness | 0.099 mm |
| Width (Metric) | 100 mm |
| Temperature Range (Metric) | -40°C to 110°C |
| Shape | Diamond |
| Show More |