Learn More
Brady™ GHS Symbol - Flammable, Card Mounted
Product Code. 15494841 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
100 x 100 mm
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15494841 100 x 100 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15494841 Supplier Brady™ Supplier No. 811683
  • ONLINE EXCLUSIVE PRICE
    193.00 NOK
    187.10 NOK / Pack of 4
    Save 5.90 NOK
    3% Off
Estimated Shipment: 24-11-2026
Pre-printed labels to identify your chemical product containers.
  • High-quality labels that can withstand the most demanding environments
  • Highly resistant to bad weather, abrasion and chemical products
Specifications
Alternate Part Number PIC 1802-100*100-B7541-CRD
Finish Gloss
Material Polyester
Quantity 4/Pk.
Adhesive Type Permanent Acrylic
Height (Metric) 100 mm
Thickness 0.099 mm
Width (Metric) 100 mm
Temperature Range (Metric) -40°C to 110°C
Shape Diamond
Dimensions (L x W) 100 x 100 mm
Color Black on White