Learn More
Brady™ GHS Symbol - Corrosive, Continuous roll
Product Code. 15694227 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
100 x 100 mm
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15694227 100 x 100 mm Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15694227 Supplier Brady™ Supplier No. 814030
  • ONLINE EXCLUSIVE PRICE
    4555.00 NOK
    4490.69 NOK / Pack of 250
    Save 64.31 NOK
    1% Off
In Stock
Pre-printed labels to identify your chemical product containers.
  • High-quality labels that can withstand the most demanding environments
  • Highly resistant to bad weather, abrasion and chemical products
Specifications
Alternate Part Number PIC 1808-100*100-B7541-RLL-C
Finish Gloss
Material Polyester
Quantity 250/Pk.
Adhesive Type Permanent Acrylic
Height (Metric) 100 mm
Thickness 0.099 mm
Width (Metric) 100 mm
Temperature Range (Metric) -40°C to 110°C
Shape Diamond
Dimensions (L x W) 100 x 100 mm
Color Black on White