Learn More
Brady™ GHS Symbol - Acute Toxicity, Continuous roll
Product Code. 15604237 Shop All Brady Products
Change view
Click to view available options
Material:
Polyester
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
15604237 Polyester Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15604237 Supplier Brady™ Supplier No. 814031
  • ONLINE EXCLUSIVE PRICE
    4555.00 NOK
    4490.69 NOK / Pack of 250
    Save 64.31 NOK
    1% Off
View all promotions
Estimated Shipment: 16-12-2026
Pre-printed labels to identify your chemical product containers.
  • High-quality labels that can withstand the most demanding environments
  • Highly resistant to bad weather, abrasion and chemical products
Specifications
Alternate Part Number PIC 1809-100*100-B7541-RLL-C
Finish Gloss
Material Polyester
Quantity 250/Pk.
Adhesive Type Permanent Acrylic
Height (Metric) 100 mm
Thickness 0.099 mm
Width (Metric) 100 mm
Temperature Range (Metric) -40°C to 110°C
Shape Diamond
Color Red