Learn More
Abnova™ GH2 (Human) Recombinant Protein
Product Code. 16135441
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. Quantity unitSize
16135441 10 μg 10µg
16145441 25 μg 25µg
2 options
Product Code. 16135441
Brand: Abnova™ H00002689P01.10ug
Human GH2 full-length ORF ( AAH20760.1, 27 a.a. - 217 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: FPTIPLSRLFDNAMLRARRLYQLAYDTYQEFEEAYILKEQKYSFLQNPQTSLCFSESIPTPSNRVKTQQKSNLELLRISLLLIQSWLEPVQLLRSVFANSLVYGASDSNVYRHLKDLEEGIQTLMWRLEDGSPRTGQIFNQSYSKFDTKSHNDDALLKNYGLLYCFRKDMDKVETFLRIVQCRSVEGSCGF
Specifications
Accession Number AAH20760.1
Gene ID (Entrez) 2689
Name growth hormone 2
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias GH-V, GHL, GHV, hGH-V
Gene Symbol GH2
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer