missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GGT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA586845
Additional Details : Weight : 0.01000kg
Description
Glutamyltransferase (gGT) from human kidney is a membrane-bound enzyme that transfers the gamma-glutamly moiety of gamma-glutamyl compounds such as glutathione to an acceptor, which may be an amino acid or a di- or tripeptide or water. The principal functions of gGT may be hydrolysis and metabolism of glutathione. Various human gGT isoforms have been found. The molecular mass of gGT for normal kidney is 90kDa and gGT can be used as a renal cell marker.Specifications
GGT1 | |
Polyclonal | |
Unconjugated | |
GGT1 | |
Cathepsin L1; Cathepsin L; Major excreted protein; MEP; CTSL; CTSL1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
116568, 14598, 2678 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P07314, P19440, Q60928 | |
Ggt1 | |
A synthetic peptide corresponding to residues in Human GGT1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |