missing translation for 'onlineSavingsMsg'
Learn More

GGPS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16893864 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16893864 150 μL 150µL
16803874 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16893864

Brand: Proteintech 149441AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene is a member of the prenyltransferase family and encodes a protein with geranylgeranyl diphosphate (GGPP) synthase activity. The enzyme catalyzes the synthesis of GGPP from farnesyl diphosphate and isopentenyl diphosphate. GGPP is an important molecule responsible for the C20-prenylation of proteins and for the regulation of a nuclear hormone receptor. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

Specifications

Antigen GGPS1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GGPS1
Gene Accession No. O95749, Q6F596, Q9WTN0
Gene Alias GGPP synthase, GGPPS, GGPPS1, GGPPSase, GGPS1
Gene Symbols Ggps1
Host Species Rabbit
Immunogen GGPS1 Fusion Protein Ag6745
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14593, 291211, 9453
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.