Learn More
Thermo Scientific™ GFP-STAT3
Description
STAT3 expressed as a fusion with GFP (Green Fluorescent Protein) to act as a physiological substrate for the Janus family of tyrosine kinases.
For use in combination with the LanthaScreen™ Tb-anti-STAT3 [pTyr705] Antibody (Cat. No. PV4846) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Specifications
| Chromagen Quantity | 10nmol |
| Content And Storage | GFP-STAT3 protein substrate in 50 mM Tris-HCl pH 7.5, 100 mM NaCl, 10% Glycerol, 1 mM DTT Store at -80°C. |
| Detection Method | Fluorescence |
| For Use With (Application) | Pharma and biopharma, target and lead identification and validation, combination with the LanthaScreen™ Tb-anti-STAT3 [pTyr705] Antibody (PV4846) to evaluate inhibitors in medium to high throughput screening applications, drug discovery and development, kinase biology, biochemical kinase assays, LanthaScreen™ kinase activity assays |
| Quantity | 10 nmol |
| Substrate | STAT3 |
| Substrate Properties | Protein-Based Substrate |
| Substrate Type | Kinase Substrate |
| Target Enzyme | Tyrosine Kinases |
| Item Description | GFP-STAT3, 10nmol, Expressed as a Fusion with GFP (Green Fluorescent Protein) to act as a Physiological Substrate For the Janus Family of Tyrosine Kinases |
| Show More |