missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Antibody (1025527) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB42402JF525
This item is not returnable.
View return policy
Description
GFP Monoclonal antibody specifically detects GFP in Broad Species Reactivity samples. It is validated for Western Blot,Simple Western,Flow Cytometry (Intracellular Staining),Immunocytochemistry
Specifications
| GFP | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| E. coli-derived recombinant GFP, aa 2-238 | |
| RUO | |
| Broad Species Reactivity | |
| Liquid |
| Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
| 1025527 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction