Learn More
Geranylgeraniol (Synthetic), TRC
Specifications
Specifications
| Chemical Name or Material | Geranylgeraniol (mixture of isomers) |
| CAS | 24034-73-9 |
| Molecular Formula | C20 H34 O |
| Quantity | 50 mg |
| Synonym | (2E,6E,10E)-3,7,11,15-Tetramethyl-2,6,10,14-hexadecatetraen-1-ol, (E,E,E)- 3,7,11,15-Tetramethyl-2,6,10,14-hexadecatetraen-1-ol, (E,E,E)-Geranylgeraniol, All-trans-Geranylgeraniol, trans,trans,trans-Geranylgeraniol, trans-Geranylgeraniol |
| InChI Formula | InChI=1S/C20H34O/c1-17(2)9-6-10-18(3)11-7-12-19(4)13-8-14-20(5)15-16-21/h9,11,13,15,21H,6-8,10,12,14,16H2,1-5H3/b18-11+,19-13+,20-15+ |
| SMILES | CC(=CCC\C(=C\CC\C(=C\CC\C(=C\CO)\C)\C)\C)C |
| IUPAC Name | (2E,6E,10E)-3,7,11,15-tetramethylhexadeca-2,6,10,14-tetraen-1-ol |
| Molecular Weight (g/mol) | 290.48 |
| Formula Weight | 290.261 |