Learn More
Gibco™ Gentamicin (10 mg/mL)
Descripción
Gentamicin sulfate is a water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea. Gentamicin acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition protein synthesis and death in susceptible bacteria. Gibco™ Gentamicin is effective against a wide variety of gram-positive and gram-negative bacteria, and is used for the prevention of bacterial contamination on cell cultures. The recommended working concentration ranges from 0.5 to 50μg/ml. We offer a variety of antibiotics and antimycotics for cell culture applications.
Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
Dual-Site cGMP Manufacturing
Gibco™ Gentamicin is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Gentamicin product made in our Grand Island facility (15710-072). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
Dual-Site cGMP Manufacturing
Gibco™ Gentamicin is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Gentamicin product made in our Grand Island facility (15710-072). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Especificaciones
Especificaciones
| Certifications/Compliance | ISO 13485, cGMP, FDA |
| Content And Storage | 15 to 30°C |
| Description | Gentamicin, 20 x 100mL, 10mg/mL, 15 to 30°C, Storage Temperature, Antibiotic, Gentamicin Agent, Antibiotic Reagent Type, 24 Months Shelf Life, ISO 13485/cGMP/FDA Certified, Ambient, Fewer Resources Used/Less Waste |
| Concentration | 10 mg/mL |
| Form | Liquid |
| Product Type | Antibiotic |
| For Use With (Application) | Mammalian cell culture, prevention of cell culture contamination |
| Quantity | 20 x 100mL |
For Research Use Only. Not for use in diagnostic procedures.
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?