Learn More
Genevac™ Refrigerated MiVac SpeedTrap
Specifications
Specifications
| Type | Refrigerated MiVac SpeedTrap |
| For Use With (Equipment) | Solvent trap |
| Capacity | 2 L |
| Length (Metric) | 212 mm |
| Height (Metric) | 450 mm |
| Voltage | 230 V |