Learn More
Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer Accessories
Product Code. 15530602 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Dust Cover
Replacement Compartment Lid
Self-Stick Printer Paper
Standard Printer Paper
Thermal Snap-on Printer
Tungsten Halogen Lamp
Unit Size:
10 rolls
Each
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15530602 Tungsten Halogen Lamp Each
15560592 Thermal Snap-on Printer Each
15681465 Self-Stick Printer Paper 10 rolls
15551182 Standard Printer Paper Pack of 10
15500602 Dust Cover Each
15510602 Replacement Compartment Lid Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15530602 Supplier Thermo Scientific™ Supplier No. 840278700
  • ONLINE EXCLUSIVE PRICE
    2112.63 NOK
    1827.93 NOK / Pack of 1
    Save 284.70 NOK
    13% Off
Estimated Shipment: 12-11-2026
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
Pre-aligned, pre-assembled replacement lamp
Customer replaceable - slides into place
Mouse-hole replacement access.  Cover removal not required.
TRUSTED_SUSTAINABILITY
Specifications
Type Tungsten Halogen Lamp
For Use With (Equipment) GENESYS 30 Visible Spectrophotometer
Includes 1 pre-aligned lamp assembly