Learn More
Applied Biosystems™ GeneScan™ Installation Standard DS-33 , [6FAM™, VIC™, NED™, PET™ dyes]
Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
Description
Includes
4 Tubes of fluorescently labeled pooled PCR products, 1 tube of GeneScan -500 LIZ Size Standard, and 1 tube of loading buffer
The DS-33 GeneScan™ Installation Standards using 6FAM™, VIC™, NED™ and PET™ dyes contain pooled PCR products amplified from microsatellite loci present in CEPH individual 1347. The kit also contains a tube of GeneScan™ 500 LIZ™ Size Standard.
Order Info
Each
Specifications
Specifications
| Content And Storage | This kit contains 4 tubes of fluorescently labeled pooled PCR products and 1 tube of GeneScan 500 LIZ Size Standard. Store the kit at -15 to -25°C until ready to use. Upon thawing, store the GeneScan 500 LIZ Size Standard tube at 2–8°C. |
| For Use With (Equipment) | 3730xl DNA Analyzer, 3730 DNA Analyzer |
| Product Line | FAM, NED PET, VIC |
| Type | Installation Standard |
| Label or Dye | FAM, NED, PET, VIC |
| Quantity | 1 kit |
| Shipping Condition | Dry Ice |