Learn More
3M™ General Purpose Vinyl Tape 764i
Product Code. 17217780
Change view
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
Unit Size:
24 rolls
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17217780 Black 24 rolls
17227780 Blue 24 rolls
17237780 Red 24 rolls
17247780 Yellow 24 rolls
17257780 White 24 rolls
17267780 Green 24 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17217780 Supplier 3M™ Supplier No. 7000048917
  • ONLINE EXCLUSIVE PRICE
    3945.00 NOK
    2354.26 NOK / 24 rolls
    Save 1590.74 NOK
    40% Off
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
Used for all non-critical applications, including colour coding, bundling and safety marking. Our non-corrosive, flexible vinyl tape is ideal for protecting parts, equipment and products from abrasions.
  • Use for taping in all non-critical applications
  • Protects parts, equipment and products from abrasions
  • Bundle items, bundle and temporarily mark parts, machines, plus much more
  • Maintains vivid colour over time for long-lasting quality
  • Can be applied by hand or with floor applicator
  • Ideal for marking applications in light traffic areas. Lots of different colours available such as yellow, red, blue, green, black, white, etc
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W) 50 mm x 33 m
Length (Metric) 33 m
Thickness 0.13 mm
Width (Metric) 50 mm
Color Black