Learn More
3M™ General Purpose Vinyl Tape 764
Product Code. 17791689 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
Unit Size:
12 rolls
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17791689 Blue 12 rolls
17771689 White 12 rolls
17781689 Red 12 rolls
17761689 Yellow 12 rolls
17721689 Green 12 rolls
17721699 Black 12 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17791689 Supplier 3M™ Supplier No. 7100260288
Estimated Shipment: 14-01-2027
This vinyl tape is a cost-effective choice for a variety of hazard, safety marking, and color-coding applications when you’re not dealing with tough conditions or the need for a long-term clean removal. Comes individually wrapped and conveniently packaged.
  • Ideal for marking applications in light traffic areas, walls and pipes
  • Use for taping in all non-critical applications
  • Maintains vivid color even when exposed to abrasion and solvents
  • Protects parts, equipment and products from abrasions
  • Bundle and temporarily mark parts, machines, and more
  • Easy-to-apply tape stretches around corners for a reliable, tight fit
  • Fast tack and high holding strength
  • Available in wide variety of colors
  • Can be applied by hand or with floor applicator
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W) 33 m x 50 mm
Packaging Quantity 12 Rolls
Thickness 0.12 mm
Adhesive Type Rubber-based Adhesive
Color Blue
Material Vinyl