Learn More
Buerkle™ General Purpose Aseptic Polypropylene Sample Box
Shop All Bürkle Products
Click to view available options
Diameter (Metric):
31 mm
43 mm
75 mm
Capacity (Metric):
50 mL
90 mL
300 mL
Height (Metric):
78 mm
80 mm
88 mm
Unit Size:
Each
Description
- PP, transparent
- With area for labelling and with graduation
- Liquid/condensate in the lid runs into an absorbing groove integrated in the lid, nothing is lost, nothing can drip
- The box is completely tight, has no crevices, is therefore easy to fill and can be emptied without residues
- Produced under aseptic conditions
- Autoclavable and suitable for deep freezing
Specifications
Specifications
| Color | Translucent |
| Diameter (Metric) | 31 mm |
| Product Type | Sample Box |
| Description | Aseptic Sample box |
| Closure Color | Transparent |
| Autoclavable | Autoclavable |
| Capacity (Metric) | 50 mL |
| Material | Polypropylene |
| For Use With (Application) | Suitable for Use with Foodstuffs |
| Includes | Sealing cover |
| Show More |