Learn More
Corning™ General Assay Microplate Lids
GREENER_CHOICE
Product Code. p-152205 Shop All Corning Products
Click to view available options
Plate Compatibility:
96-well Microplates Only (not 2 mL block)
96-well Plate
All Microplates
No. per Case:
100
100/Cs.
50/Cs.
Material:
Acetate
Polystyrene
Unit Size:
Pack of 100
Pack of 50
Product Code. 10689321
Brand: Corning™ 3931
Only null left
Add to basket
Add to basket
Manufactured with rigid polystyrene for optimum visibility and strength. Corning™ General Assay Microplate Lids are available in a variety of styles and types.
  • All lids are made of rigid polystyrene except where indicated.
  • All lids have a corner notch on the A1 corner (except where indicated) to correspond to the corner notches found on all Corning™ microplates.
  • The universal lid without a corner notch (Cat. No. 3098) does not need to be oriented in any particular direction to be placed on Corning microplates. The lid also has a shorter skirt than standard lids.
  • The black universal lid (Cat. No. 3935) is suitable for fluorescent and other light-sensitive assays.
  • The DMSO-resistant cyclic olefin copolymer (COC) lid (Cat. No. 3085) is tinted amber in color for light-sensitive assays and is 100% DMSO-resistant.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Description Low evaporation lid with corner notch and condensation rings
    Material Polystyrene
    Sterility Sterile
    Condensation Rings Yes
    No. per Case 50/Cs.
    No. per Pack 25/Pk.
    Product Type Lid
    Plate Compatibility 96-well Microplates Only (not 2 mL block)
    Cut-off Corners Corner Notch