Learn More
MP Biomedicals™ GeneClean Kit
Product Code. 11363099 Shop All MP Biomedicals Products
Change view
Click to view available options
No. of Reactions:
200 Preps
Unit Size:
200 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
11363099 200 Preps 200 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11363099 Supplier MP Biomedicals™ Supplier No. 111001200
  • ONLINE EXCLUSIVE PRICE
    3500.00 NOK
    3331.61 NOK / Pack of 200
    Save 168.39 NOK
    4% Off
 Restricted Item
Estimated Shipment: 14-12-2026
GeneClean™ Kit, Solution or Gels
  • The GENECLEAN™ Kit includes Glassmilk™ slurry that allows flexibility of reaction size and volume.
  • The GENECLEAN™ kit simplifies the process of purifying DNA into three easy steps: Bind,Wash and Elute.
  • Ethanol precipitation is never required and purified DNA is immediately ready for a wide variety of downstream applications.
  • Isolate DNA from TAE buffered agarose gels and solutions
  • The GENECLEAN™ Kit is used to purify fragments of DNA from 200 bp to 20 kb from solutions and agarose gels.
  • Examples of applications are desalting, elimination of proteins from enzymatic reactions, removal of primers and unincorporated nucleotides from enzymatic reactions, separation of linearized from uncut vector and isolation of PCR products away from genomic DNA.
Specifications
Concentration New Wash/Ethanol Concentration must be ∽ 50%
Content And Storage Store at Room Temperature (15-30°C). NaI solution must be kept away from light and open air sources to prevent oxidation. NaI is packaged under nitrogengas to further reduce chance of oxidation. Shelf life may be extended by storing at 4°C.
For Use With (Application) Examples of applications are desalting, elimination of proteins from enzymatic reactions, removal of primers and unincorporated nucleotides from enzymatic reactions, separation of linearized from uncut vector and isolation of PCR products away fromgenomic DNA.
No. of Reactions 200 Preps
pH Above 500 bp: 4.5 M NaI pH 7.0 - 7.4; Between 200 - 500 bp: 4.5 M NaI pH 6.0; NEW Wash: pH Range: 7.4 - 8.0
Product Type Kits and Assays
Form Solution or Gels