Learn More
Thermo Scientific™ GeneBLAzer™ PPAR delta DA Assay Kit
Product Code. 10073664
Click to view available options
Quantity:
1 plate
Unit Size:
Each
Product Code. 10073664
Brand: Thermo Scientific™ K1395
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
GeneBLAzer™ PPAR delta DA (Division Arrested) cells and PPAR delta-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Peroxisome Proliferator-Activated Receptor-delta (PPAR delta) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™ UAS-bla HEK 293T cell line.
GeneBLAzer™ PPAR delta DA (Division Arrested) cells and PPAR delta-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Peroxisome Proliferator-Activated Receptor-delta (PPAR delta) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™ UAS-bla HEK 293T cell line. GeneBLAzer™ UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-PPAR delta (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both PPAR delta DA cells and PPAR delta-UAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of L-165041. In addition, PPAR delta-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.
TRUSTED_SUSTAINABILITY
Specifications
Alternate Agonists Estrone, Genistein, Diethylstibestrol, Ethynylestradiol
Antagonists 4-OH-tam+0.2 nM E2, Tam+0.2 nM E2, 4-OH-tamoxifen, Tamoxifen, RU486+0.2 nM E2, RU486
Assay Entry Cell-based beta lactamase reporter gene
Cell State Division-Arrested Cells
Content And Storage Contains sufficient division-arrested cells and substrate to assay 1 x 384-well plate:
• PPAR delta HEK 293T DA cells (K1395A)
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg
Shipping: Dry Ice
Storage: -80°C (liquid nitrogen)
Description GeneBLAzer™ PPAR delta DA Assay Kit, 1 Plate, ∽4,800,000 cells/vial at 4,800,000 cells/mL Quantity of Cells, 293T (HEK) Cell Line, Fluorescent Detection Method, Nuclear Receptors Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Detection Method Fluorescence
For Use With (Application) Cellular imaging, pharma and biopharma, target and lead identification and validation, target-based adme/tox assays, Cell analysis, cell-based reporter assays, cell-based and lactamase assays, drug discovery and development, nuclear receptor biology, cell-based nuclear receptor reporter assays, target-based ADME/tox assays, cellular adme assays, nuclear receptor cellular adme assays
Gene ID (Entrez) 5467
Gene Symbol PPARδ
Includes PPAR delta HEK 293T DA cells, LiveBLAzer™-FRET B/G Loading Kit, 70μg
Primary Agonist 17-β-estradiol
Product Line GeneBLAzer
Quantity 1 plate
Reporter Gene BLA (Beta-Lactamase)
Sample Type 293T (HEK)
Target Entry PPAR delta, NR1C2, PPARD
Type Division-Arrested Cells Assay Kit
Cell Line 293T (HEK)
System Type GeneBLAzer™
Druggable Target Nuclear Receptors