Learn More
Thermo Scientific™ GeneBLAzer™ P2RY4-NFAT-bla CHO-K1 Cells
Product Code. 10535854
Change view
Click to view available options
Quantity:
1 vial
Unit Size:
Each
Product Code. Quantity unitSize
10535854 1 vial Each
1 options
Product Code. 10535854
Brand: Thermo Scientific™ K1863
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
GeneBLAzer™ P2RY4-NFAT-bla CHO-K1 cells contain the human Purinergic Receptor P2Y4 (P2RY4), stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line.
GeneBLAzer™ P2RY4-NFAT-bla CHO-K1 cells contain the human Purinergic Receptor P2Y4 (P2RY4), stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1078) contain a beta-lactamase reporter gene under control of the NFAT Response Element. The GeneBLAzer™ P2RY4-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of UTP. In addition, GeneBLAzer™ P2RY4-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number NM_002565.3
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage GeneBLAzer™ P2RY4-NFAT-bla CHO-K1 cells are shipped on dry ice, 8 x 106 cells/vial. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Detection Method Fluorescence
For Use With (Application) FRET
Human Protein Class G-Protein Coupled Receptor (GPCR)
Population Status Pool of Stable Cells
Product Line GeneBLAzer
Quantity 1 vial
Readout End Point
Target Entry P2RY4
Type Dividing Cells
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR