Learn More
Thermo Scientific™ GeneBLAzer™ NPSR1-B-NFAT-bla CHO-K1 Cells
Product Code. 10494425
Click to view available options
Quantity:
2 x 106 cells
Unit Size:
Each
Product Code. 10494425
Brand: Thermo Scientific™ K1481B
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The GeneBLAzer™ NPSR1-B-NFAT-bla CHO-K1 cells contain the human Neuropeptide S Receptor 1-Isoform B (NPSR1-B), (Accession # NP_997056) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line.
CellSensor™ NFAT-bla CHO-K1 cells (Cat #K1078) contain a beta-lactamase (bla) reporter gene under control of the NFAT response element.
The NPSR1-B-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of Neuropeptide-S. In addition, NPSR1-B-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number NP_997056.1
Cell State Dividing Cells
Content And Storage (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
o GeneBLAzer™ NPSR1-B-NFAT-bla CHO-K1 cells
o GeneBLAzer™ NPSR1-B-NFAT-bla CHO-K1 Assay Protocol
o Certificate of Analysis
The GeneBLAzer™ NPSR1-B-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
Gene Symbol NPSR1-B
Quantity 2 x 106 cells
Reporter Gene BLA (Beta-Lactamase)
Type CHO-K1 Cells
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR