Learn More
Thermo Scientific™ GeneBLAzer™ GPR54-NFAT-bla CHO-K1 Cells
Product Code. 10686414
Change view
Click to view available options
Quantity:
2 x 106 Cells
Unit Size:
Each
Product Code. Quantity unitSize
10686414 2 x 106 Cells Each
1 options
Product Code. 10686414
Brand: Thermo Scientific™ K1720
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
GeneBLAzer™ GPR54-NFAT-bla CHO-K1 cells contain the human KiSS1 receptor (GPR54) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line.
GeneBLAzer™ GPR54-NFAT-bla CHO-K1 cells contain the human KiSS1 receptor (GPR54) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1534) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element.
The GeneBLAzer™ GPR54-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of Metastin. In addition, GeneBLAzer™ GPR54-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
The GPR54 receptor has been shown to be involved in tumor progression and metastasis, and in the development and functional regulation of the gonads.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number NP_115940.1
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.
Includes:
• GeneBLAzer™ GPR54-NFAT-bla CHO-K1 cells
• GeneBLAzer™ GPR54-NFAT-bla CHO-K1 Assay Protocol
• Certificate of Analysis
The GeneBLAzer™ GPR54-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Description GeneBLAzer™ GPR54-NFAT-bla CHO-K1 Cells, 2 x 106 Cells, ∽2,600,000 cells/vial at 2,600,000 cells/mL Quantity of Cells, CHO-K1 Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol GPR54
Includes GeneBLAzer™ GPR54-NFAT-bla CHO-K1 cells (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). Includes: GeneBLAzer™ GPR54-NFAT-bla CHO-K1 cells, GeneBLAzer™ GPR54-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Sample Type CHO-K1
Target Entry GPR54
Type CHO-K1 Cells
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR