Learn More
Thermo Scientific™ GeneBLAzer™ EDG7-NFAT-bla HEK 293T Cells
Code produit. 10267413
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
2 x 106 Cells
missing translation for 'unitSize'
Each
Code produit. Quantity unitSize
10267413 2 x 106 Cells Each
1 missing translation for 'options'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 10267413
missing translation for 'mfr': Thermo Scientific™ K1511
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
GeneBLAzer™ EDG7-NFAT-bla HEK293T cells contain the human Endothelial Differentiation Gene (EDG7) stably integrated into the CellSensor™ NFAT-bla HEK293T cell line.
GeneBLAzer™ EDG7-NFAT-bla HEK293T cells contain the human Endothelial Differentiation Gene (EDG7) stably integrated into the CellSensor™ NFAT-bla HEK293T cell line. CellSensor™ NFAT-bla HEK293T cells (Cat. No. K1538) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element.
The GeneBLAzer™ EDG7-NFAT-bla HEK293T cells are functionally validated for Z' and EC50 concentrations of 18:1 LPA. In addition, GeneBLAzer™ EDG7-NFAT-bla HEK293T cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
LPA receptors were originally classified in the Endothelial Differentiation Gene (EDG) family of receptors. The EDG family was later divided into two distinct groups of receptors based on their ligand specificity for either Lysophosphatidic acid (LPA) or Sphingosine-1-Phosphate (S1P). There are three high affinity LPA receptors LPA1, LPA2, and LPA3 (formally EDG2, EDG4, and EDG7), and five high affinity S1P receptors S1P1, S1P2, S1P3, S1P4 and S1P5 (formally EDG1, EDG5, EDG3, EDG6, and EDG8). LPA receptors are differentially linked via Gi and Gq to multiple effector systems and diverse biological functions. Recent studies revealed that targeted deletion of the EDG7/LPA3 receptor results in delayed implantation and altered embryo spacing, and significantly reduced litter size in mice. These findings indicate that EDG7 plays a crucial role in implantation and mammalian reproduction.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
TRUSTED_SUSTAINABILITY
Spécification
Accession Number NM_012152.1
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.
Includes:
• GeneBLAzer™ EDG7-NFAT-bla HEK293T cells
• GeneBLAzer™ EDG7-NFAT-bla HEK293T Assay Protocol
• Certificate of Analysis
Storage:
The GeneBLAzer™ EDG7-NFAT-bla HEK293T cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol EDG7
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Target Entry EDG7
Cell Line 293T (HEK)
System Type GeneBLAzer™
Druggable Target GPCR
Afficher plus Afficher moins
For Research Use Only. Not for use in diagnostic procedures.
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.