Learn More
Thermo Scientific™ GeneBLAzer™ C5AR1-Gα15 CHO-K1 DA Assay Kit
Description
The GeneBLAzer™ C5AR1-Gα15 CHO-K1 DA (Division Arrested) cells contain the human Complement Component 5a Receptor 1 (C5AR1) (Accession No. NM_000710) stably integrated into the GeneBLAzer™ Gα15-NFAT-bla CHO-K1 cell line. GeneBLAzer™ Gα15-NFAT-bla CHO-K1 (Cat. No. K1213) contains a beta-lactamase (bla) reporter gene under control of a NFAT response element and a promiscuous G Protein, Gα15, stably integrated into CHO-K1 cells.
The C5AR1-Gα15 CHO-K1 DA (Division Arrested) cells cells are functionally validated for Z'-factor and EC50 concentrations using C5a. In addition, C5AR1-Gα15-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin C, and have no apparent toxicity or change in cellular signal transduction.
Specifications
Specifications
| Accession Number | NM_001736.2 |
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Division-Arrested Cells |
| Content And Storage | Each kit contains sufficient quantities of division arrested (DA) cells and substrate to assay 1 x 384 well plate. (Other materials are required separately; please refer to the protocol). Includes: • GeneBLAzer™ C5AR1-Gα15 CHO-K1DA cells • LiveBLAzer™-FRET B/G Loading Kit, 70 μg • Solution D • GeneBLAzer™ C5AR1-Gα15 CHO-K1 Assay Protocol • LiveBLAzer™-FRET B/G Loading Protocol • Certificate of Analysis The GeneBLAzer™ C5AR1-Gα15 CHO-K1 DA cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
| Description | GeneBLAzer™ C5AR1-Gα±15 CHO-K1 DA Assay Kit, 1 Plate, ∽4,800,000 cells/vial Quantity of Cells, CHO-K1 Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, GeneBLAzer™ System, Liquid Nitrogen Storage |
| Detection Method | Fluorescence |
| For Use With (Application) | Pharma and biopharma, drug discovery and development, target and lead identification and validation, G-protein coupled receptor biology, cell-based GPCR reporter assays |
| Gene Symbol | C5aR1 |
| Includes | Each kit contains sufficient quantities of division arrested (DA) cells and substrate to assay 1 x 384 well plate. (Other materials are required separately; please refer to the protocol). Includes: GeneBLAzer™ C5AR1-G?15 CHO-K1DA cells, LiveBLAzer™-FRET B/G Loading Kit, 70μg, Solution D, GeneBLAzer™ C5AR1-G?15 CHO-K1Assay Protocol, LiveBLAzer™-FRET B/G Loading Protocol, Certificate of Analysis |
| Population Status | Single Cell Clone Origin |
| Show More |