missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech GEN1 Mouse anti-Human, Clone: 1A10D10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68060-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
GEN1 Monoclonal antibody apecifically detetects GEN1 antigen in Human Samples. It is validated for Western Blot, ELISA.Specifications
GEN1 | |
Monoclonal | |
1000 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
GEN1 | |
GEN1 fusion protein Ag31247 | |
20 μL | |
348654 | |
Store at -20°C. Stable for one year after shipment. | |
IgG1 |
Western Blot, ELISA | |
1A10D10 | |
Unconjugated | |
NM_182625 | |
Mouse | |
Protein G purification | |
Primary | |
Human | |
Liquid |