Learn More
Gemfibrozil Ethyl Ester, Mikromol™
Product Code. 17933356
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 17933356
Brand: Mikromol MM0641.14
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Gemfibrozil Ethyl Ester
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Gemfibrozil Ethyl Ester
CAS 63258-18-4
Analyte or Component Names Gemfibrozil Ethyl Ester
Shipping Condition Room Temperature
Molecular Formula C17 H26 O3
Certifications/Compliance ISO 9001
Synonym Gemfibrozil Ethyl Ester, 5-(2,5-Dimethylphenoxy)-2,2-dimethyl-pentanoic acid ethyl ester
InChI Formula InChI=1S/C17H26O3/c1-6-19-16(18)17(4,5)10-7-11-20-15-12-13(2)8-9-14(15)3/h8-9,12H,6-7,10-11H2,1-5H3
SMILES CCOC(=O)C(C)(C)CCCOc1cc(C)ccc1C
IUPAC Name ethyl 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoate
Molecular Weight (g/mol) 278.39
Quantity 100 mg
Formula Weight 278.1882 g/mol