Learn More
Cytiva GelBond PAG Gel Film
Shop All Cytiva Products
Click to view available options
For Use With (Application):
Agarose gels
Polyacrylamide gels
Length (Metric):
94 mm
258 mm
260 mm
Width (Metric):
84 mm
124 mm
203 mm
Unit Size:
Pack of 50
Description
Cytiva™ GelBond Film is a transparent, flexible polyester film designed to support gels. Gels cast on GelBond Film remain permanently attached to the film through electrophoresis or immunodiffusion and all subsequent fixing, staining, destaining, and drying procedures.
- GelBond Film is made from thin polyester (0.2mm). Pack of 50 films, 124 x 258mm.
- High-density homogenous medium
Specifications
Specifications
| Product Type | Gel Film |
| For Use With (Application) | Polyacrylamide gels |
| Quantity | 50 Pack |
| Length (English) | 10.2 in. |
| Length (Metric) | 260 mm |
| Width (English) | 7.9 in. |
| Width (Metric) | 203 mm |