Learn More
Cytiva GelBond PAG Gel Film
Product Code. 10086234 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Application):
Agarose gels
Polyacrylamide gels
Length (Metric):
94 mm
258 mm
260 mm
Width (Metric):
84 mm
124 mm
203 mm
Unit Size:
Pack of 50
Product Code. For Use With (Application) Length (Metric) Width (Metric) unitSize
10086234 Polyacrylamide gels 258 mm 124 mm Pack of 50
11520824 Polyacrylamide gels 258 mm 124 mm Pack of 50
10075804 Agarose gels 258 mm 124 mm Pack of 50
10575785 Agarose gels 94 mm 84 mm Pack of 50
11530824 Polyacrylamide gels 260 mm 203 mm Pack of 50
10754037 Polyacrylamide gels 260 mm 203 mm Pack of 50
6 options
Product Code. 10086234
Brand: Cytiva 80112936
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Cytiva™ GelBond PAG Gel Film is a high density homogenous medium.
Cytiva™ GelBond Film is a transparent, flexible polyester film designed to support gels. Gels cast on GelBond Film remain permanently attached to the film through electrophoresis or immunodiffusion and all subsequent fixing, staining, destaining, and drying procedures.
  • GelBond Film is made from thin polyester (0.2mm). Pack of 50 films, 124 x 258mm.
  • High-density homogenous medium
Each film has one hydrophilic side and one hydrophobic side. The gel must be cast on the hydrophilic side, which is the side where a water droplet will spread out.
TRUSTED_SUSTAINABILITY
Specifications
Color Colorless
For Use With (Application) Polyacrylamide gels
Quantity 50 Pack
Length (English) 10.1 in.
Length (Metric) 258 mm
Width (English) 5.0 in.
Width (Metric) 124 mm
Product Type Gel Film