Learn More
Lonza™ GelBond™ Film for Agarose Gels
Product Code. 11597755
Click to view available options
Length (Metric):
12.5 cm
15 cm
16.5 m
18 cm
20.5 cm
24.5 cm
25.8 cm
Width (Metric):
10 cm
10.2 cm
11 cm
12.4 cm
12.5 cm
12.7 cm
16 cm
20.3 cm
Unit Size:
1 roll
100 sheets
Pack of 100
Product Code. 11597755
Brand: Lonza™ 53780
Transparent, flexible polyester film supports gels through electrophoresis, immunodiffusion, fixing, staining, destaining, and drying procedures. GelBond™ Film for Agarose Gels are reliable, durable and convenient.
  • Permanently attaches to gels during casting
  • Makes gels easier to handle
  • Helps gels retain their original dimensions during staining and drying
  • Available as precut sheets or rolls
Note: polyester films will not transmit light of less than 310nm and will fluoresce at higher wavelengths
TRUSTED_SUSTAINABILITY
Specifications
Product Type Agarose Gel Film
For Use With (Application) Agarose Gel electrophoresis, immunodiffusion, fixing, staining, destaining and drying procedures
Material Polyester
Quantity 1 Roll
Thickness (English) 0.007 in.
Thickness (Metric) 0.02 cm
Length (English) 649.6 in.
Length (Metric) 16.5 m
Width (English) 7.9 in.
Width (Metric) 20.3 cm