Learn More
Cytiva Gel Staining Tray Packs
Product Code. 10794977 Shop All Cytiva Products
Change view
Click to view available options
Type:
Gasket
Glass Plate Divider
For Use With (Equipment):
Casting Stand
Hoefer SE 600 Vertical Unit
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) Unit Size
10794977 Glass Plate Divider Hoefer SE 600 Vertical Unit Each
10117194 Gasket Casting Stand Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10794977 Supplier Cytiva Supplier No. SE6102D
  • ONLINE EXCLUSIVE PRICE
    2080.00 NOK
    1960.00 NOK / Pack of 1
    Save 120.00 NOK
    5% Off
Estimated Shipment: 13-01-2027
For use with Hoefer Processor Plus fluid delivery system.
  • Club sandwich divider
  • Notched
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Pair two gel sandwiches so that up to four gels can be run at one time. 
For Use With (Equipment) Hoefer SE 600 Vertical Unit
Type Glass Plate Divider
Dimensions (L x W) 18 x 16 cm