Learn More
Cytiva Gel Processing Magnet
Product Code. 11773763 Shop All Cytiva Products
Change view
Click to view available options
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
11773763 Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11773763 Supplier Cytiva Supplier No. 80634220
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Prevents gels from sliding over one another.
  • When placed on gel staining tray, magnets prevent gels from sliding over one another
  • Prevents gel from coming in contact with splash guard
  • When placed on corners of gels with plastic backing, magnet helps keep gels submerged
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Prevent gels from sliding over one another, prevents gel from coming in contact with splash guard, keep gels submerged
Type Gel Processing Magnet
Quantity 4/Pk.