Description
- When placed on gel staining tray, magnets prevent gels from sliding over one another
- Prevents gel from coming in contact with splash guard
- When placed on corners of gels with plastic backing, magnet helps keep gels submerged
Specifications
Specifications
| For Use With (Application) | Prevent gels from sliding over one another, prevents gel from coming in contact with splash guard, keep gels submerged |
| Type | Gel Processing Magnet |
| Quantity | 4/Pk. |