Learn More
GEFT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17028-1-AP-20UL
This item is not returnable.
View return policy
Description
The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange. The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange (Souchet et al., 2002 ).Specifications
| GEFT | |
| Polyclonal | |
| Unconjugated | |
| ARHGEF25 | |
| ARHGEF25, GEFT, p63RhoGEF, RhoA/RAC/CDC42 exchange factor | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 115557, 314904, 52666 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6P720, Q86VW2, Q9CWR0 | |
| Arhgef25 | |
| GEFT Fusion Protein Ag10436 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |