Learn More
GDPD5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25703-1-AP-150UL
 View more versions of this product
Product Code. 16834265
  • 4836.10 NOK / 150µL
Description
The glycerophosphodiester phosphodiesterase domain containing 5 (GDPD5) protein, also known as GDE2, is a seven transmembrane, widely expressed protein that is necessary for spinal motor neuron differentiation and retinoid-induced neuronal outgrowth. Altered choline phospholipid metabolism is a hallmark of cancer, and the elevated expression of GDPD5 correlates with maligt choline phospholipid metabolite profiles in human breast cancer.
Specifications
Specifications
GDPD5
Polyclonal
Unconjugated
GDPD5
GDE2, GDPD5, PP1665, PP9363, UNQ1850/PRO3580
Rabbit
Antigen Affinity Chromatography
RUO
81544
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8WTR4
GDPD5
GDPD5 Fusion Protein Ag22530