missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDPD5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25703-1-AP-20UL
This item is not returnable.
View return policy
Description
The glycerophosphodiester phosphodiesterase domain containing 5 (GDPD5) protein, also known as GDE2, is a seven transmembrane, widely expressed protein that is necessary for spinal motor neuron differentiation and retinoid-induced neuronal outgrowth. Altered choline phospholipid metabolism is a hallmark of cancer, and the elevated expression of GDPD5 correlates with maligt choline phospholipid metabolite profiles in human breast cancer.Specifications
| GDPD5 | |
| Polyclonal | |
| Unconjugated | |
| GDPD5 | |
| GDE2, GDPD5, PP1665, PP9363, UNQ1850/PRO3580 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 81544 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8WTR4 | |
| GDPD5 | |
| GDPD5 Fusion Protein Ag22530 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction