missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GDI1 (Human) Recombinant Protein
Human GDI1 full-length ORF ( AAH00317, 1 a.a. - 447 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002664-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MDEEYDVIVLGTGLTECIPSGIMSVNGKKVLHMDRNPYYGGESSSITPLEELYKRFQLLEGPPESMGRGRDWNVDLIPKFLMANGQLVKMLLYTEVTRYLDFKVVEGSFVYKGGKIYKVPSTETEALASNLMGMFEKRRFRKFLVFVANFDENDPKTFEGVDPQTTSMRDVYRKFDLGQDVIDFTGHALALYRTDDYLDQPCLETVNRIKLYSESLARYGKSPYLYPLYGLGELPQGFARLSAIYGGTYMLNKPVDDIIMENGKVVGVKSEGEVARCKQLICDPSYIPDRVRKAGQVIRIICILSHPIKNTNDANSCQIIIPQNQVNRKSDIYVCMISYAHNVAAQGKYIAIASTTVETTDPEKEVEPALELLGPIDQKFVAISDLYEPIDDGCESQVFCSCSYDATTHFETTCNDIKDIYKRMAGTAFDFENMKRKQNDVFGEAEQ
Specifications
AAH00317 | |
GDP dissociation inhibitor 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GDI1 | |
GST |
2664 | |
Wheat germ expression system | |
25 μg | |
FLJ41411, GDIL, MRX41, MRX48, OPHN2, RABGD1A, RABGDIA, XAP-4 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |