Learn More
Velp Scientifica™ GDE Enzymatic Digester
BIOPHARMA
Product Code. 12891683 Shop All Velp Products
Change view
Click to view available options
For Use With (Application):
Dietary Fiber Analysis
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
12891683 Dietary Fiber Analysis Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12891683 Supplier Velp Scientifica™ Supplier No. SA30400209
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 05-01-2027
Enzymatic digestion for dietary fiber analysis by enzymes heating. It offers non-stop sample mixing for homogeneity.
  • The analytical procedure for the determination of dietary fiber involves a series of digestions of the sample by thermostable enzymes using the enzymatic digester GDE.
  • It operates in accordance with the official AOAC method for the determination of total dietary fiber.
  • The magnetic stirrer of GDE ensures homogeneity thanks to a continuous and constant stirring of the sample while avoiding overheating that could compromise the success of enzymatic digestions.
  • Set the timer for unattended operation, you can quickly set the operating time and focus on other tasks.
  • The transparent polycarbonate tank ensures visibility and enhanced resistance up to 105 °C; whilst the use of deionized water eliminates any possible creation of limestone.
  • Outstanding temperature regulation is possible with GDE: through an intuitive digital display it is possibile to set the desired temperature, with an excellent accuracy of only ± 0.2 °C.
Specifications
For Use With (Application) Dietary Fiber Analysis