missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA579304
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
GDA is an enzyme that catalyzes the hydrolytic deamination of guanine, producing xanthine and ammonia.Specifications
GDA | |
Polyclonal | |
Unconjugated | |
GDA | |
AU015411; AW047581; Cypin; cytoplasmic PSD95 interactor; GAH; Gda; GUANASE; guanine aminase; guanine aminohydrolase; Guanine deaminase; KIAA1258; NEDASIN; p51-nedasin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14544, 83585, 9615 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 4MG trehalose and 0.05MG sodium azide | |
Q9R111, Q9WTT6, Q9Y2T3 | |
GDA | |
E. coli-derived human GDA recombinant protein (Position: E220-Q446). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |