missing translation for 'onlineSavingsMsg'
Learn More

GCOM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16858174 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16858174 20 μL 20µL
16848174 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16858174 Supplier Proteintech Supplier No. 181291AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GCOM1, also named as Myozap, Gup and Gcom, belongs to the GCOM1 family. It plays a role in cellular signaling via Rho-related GTP-binding proteins and subsequent activation of transcription factor SRF. GCOM1 has 11 isoforms with MW 44-54 kDa and 64 kDa.

Specifications

Antigen GCOM1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GCOM1
Gene Accession No. Q3UIJ9, Q5EB94
Gene Alias Gcom, GCOM1, Gcom2, GRINL1A, GRINL1A combined protein, GRINL1A complex locus, GRINL1A upstream protein, Gup, Gup1, Gup2, MYOZAP, MYZAP, POLR2M
Gene Symbols GCOM1, Myzap
Host Species Rabbit
Immunogen GCOM1 Fusion Protein Ag12708
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102371, 145781, 363091
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.