missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCNF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12712-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody specifically reacts with the 55 kDa NR6A1 protein.
This gene encodes an orphan nuclear receptor which is a member of the nuclear hormone receptor family. Its expression pattern suggests that it may be involved in neurogenesis and germ cell development. The protein can homodimerize and bind DNA, but in vivo targets have not been identified. The gene expresses at least alternatively spliced transcript variants.Specifications
| GCNF | |
| Polyclonal | |
| Unconjugated | |
| NR6A1 | |
| GCNF, GCNF1, Germ cell nuclear factor, hGCNF, hRTR, NR61, NR6A1, RTR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14536, 2649, 362125 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15406, Q64249 | |
| Nr6a1 | |
| GCNF Fusion Protein Ag3395 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction