Learn More
GCLM Mouse anti-Human, Unconjugated, Clone: 1A5A4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66808-1-IG-150UL
 View more versions of this product
Product Code. 16839025
  • 4836.10 NOK / 150µL
Description
The GCLM antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GCLM. This antibody recognizes human antigen. The GCLM antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
GCLM
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1A5A4
Unconjugated
150 μL
2730
IgG2b