Learn More
Gcase activator 1, MedChemExpress
Product Code. 16482916
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16482916
Brand: Medchem Express HY104038
MedChemExpress Gcase activator 1 is an activator of glucocerebrosidase (Gcase) extracted from patent WO 2017192841 A1.
Gcase activator 1 is an activator of glucocerebrosidase (Gcase) extracted from patent WO 2017192841 A1.
Specifications
Chemical Name or Material Gcase activator 1
Physical Form Solid
CAS 1919820-28-2
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C20H30N4O2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 50 mg/mL (139.48 mM; Need ultrasonic)
SMILES O=C(C1=C2N=C(C)C=C(C)N2N=C1)N[C@@H]3CC[C@@H](OCCCCC)CC3
Molecular Weight (g/mol) 358.48
Formula Weight 358.48
Percent Purity 98.77%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.