Learn More
- Home
- Products
- Chromatography Supplies
- Chromatography Tubing
- Trajan Scientific and Medical™ GC Tubing Fused Silica (Not Deactivated)
Trajan Scientific and Medical™ GC Tubing Fused Silica (Not Deactivated)
Product Code. 15298833
Change view
Click to view available options
Overall Length (Metric):
1 m
10 m
25 m
Diameter (Metric) Inner:
0.01 mm
0.025 mm
0.04 mm
0.05 mm
0.06 mm
0.075 mm
0.1 mm
0.11 mm
0.15 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Diameter (Metric) Outer:
0.14 mm
0.15 mm
0.17 mm
0.2 mm
0.22 mm
0.285 mm
0.363 mm
0.43 mm
0.68 mm
Unit Size:
10m
25m
Each
24 product options available for selection
| Product Code. | Overall Length (Metric) | Diameter (Metric) Inner | Diameter (Metric) Outer | Unit Size |
|---|---|---|---|---|
| 15298833 | 25 m | 0.32 mm | 0.43 mm | 25m |
| 15258823 | 10 m | 0.25 mm | 0.15 mm | 10m |
| 15258833 | 10 m | 0.15 mm | 0.285 mm | 10m |
| 15208843 | 10 m | 0.53 mm | 0.68 mm | 10m |
| 15238823 | 10 m | 0.01 mm | 0.285 mm | 10m |
| 15278833 | 25 m | 0.22 mm | 0.363 mm | 25m |
| 12750740 | 1 m | 0.025 mm | 0.363 mm | Each |
| 13184359 | 1 m | 0.05 mm | 0.363 mm | Each |
| 12760740 | 1 m | 0.075 mm | 0.363 mm | Each |
| 15288833 | 10 m | 0.32 mm | 0.43 mm | 10m |
| 15258843 | 1 m | 0.15 mm | 0.363 mm | Each |
| 15268843 | 1 m | 0.1 mm | 0.363 mm | Each |
| 15218823 | 10 m | 0.11 mm | 0.17 mm | 10m |
| 15278823 | 10 m | 0.05 mm | 0.22 mm | 10m |
| 15208833 | 10 m | 0.1 mm | 0.2 mm | 10m |
| 15228823 | 25 m | 0.11 mm | 0.17 mm | 25m |
| 15228833 | 25 m | 0.1 mm | 0.363 mm | 25m |
| 15228843 | 10 m | 0.25 mm | 0.363 mm | 10m |
| 15238833 | 10 m | 0.15 mm | 0.22 mm | 10m |
| 15248823 | 10 m | 0.25 mm | 0.285 mm | 10m |
| 15248833 | 25 m | 0.15 mm | 0.22 mm | 25m |
| 15268823 | 10 m | 0.04 mm | 0.14 mm | 10m |
| 15288823 | 10 m | 0.05 mm | 0.15 mm | 10m |
| 15298823 | 10 m | 0.06 mm | 0.22 mm | 10m |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- High quality fused silica tubing
- Tubing protected with high temperature polyimide resin
- Used in a wide range of GC, LC and biotechnology applications
Specifications
Specifications
| Type | Tubing |
| For Use With (Application) | GC, LC and Biotechnology |
| Diameter (Metric) Inner | 0.32 mm |
| Diameter (Metric) Outer | 0.43 mm |
| Overall Length (Metric) | 25 m |