Learn More
Trajan Scientific and Medical™ GC Tubing Fused Silica (Not Deactivated)
Helps in maintaining a high performance chromatography system
Brand:  Trajan Scientific and Medical™ 062473
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15248833
  • 2890.00 NOK / 25m
Estimated Shipment: 04-06-2024
Description
  • High quality fused silica tubing
  • Tubing protected with high temperature polyimide resin
  • Used in a wide range of GC, LC and biotechnology applications
Specifications
Specifications
Tubing
0.15 mm
25 m
GC, LC and Biotechnology
0.22 mm