Learn More
Thermo Scientific™ GC Tool Kit for Agilent Instruments
Product Code. 13416956 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Agilent GC-MS Instruments
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
13416956 Agilent GC-MS Instruments Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13416956 Supplier Thermo Scientific™ Supplier No. 60180786
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
 Restricted Item
Estimated Shipment: 15-10-2026
Thermo Scientific™ GC Tool Kit for Agilent Instruments includes common consumable supplies and tools.
Thermo Scientific™ GC Tool Kit for Agilent Instruments includes common consumable supplies and tools.
  • Replacements included perform like original instrument parts
  • Parts list included for easy reordering
Includes:
Capillary installation gauge, injector wrench, septum nut removal tool, open-end wrenches in 7/16 x 1/2in. and 1/2 x 9/16in. sizes.
TRUSTED_SUSTAINABILITY
Specifications
Type Capillary Tool Kit
Description GC Tool Kit for Agilent Instruments
For Use With (Application) GC
For Use With (Equipment) Agilent GC-MS Instruments